Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIOshell™ A160 Peptide Phenyl-Hexyl, 2.7 μm HPLC Guard Cartridge, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022306 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Diameter (Metric) Inner:
2.1 mm
3 mm
4.6 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner
11-102-2306 4.6 mm
11-102-2304 2.1 mm
11-102-2305 3 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-102-2306 Supplier MilliporeSigma Supelco Supplier No. 577543U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The target analytes include peptides and polypeptides < 20 kDa.

  • High-speed, high-performance liquid chromatography column based on a 160 Å Fused-Core™ particle design.
  • The particle provides a thin porous shell of high-purity silica surrounding a solid silica core.
  • This particle design exhibits very high column efficiency for high molecular weight solutes (up to 20 kDa) due to the shallow diffusion paths in the 0.5 μm thick porous shell and the small overall particle size of 2.7 μm.
  • The end-capped, phenyl-hexyl bonded phase of the Peptide Phenyl-Hexyl provides a stable, reversed phase packing with a pore structure and pore size that is optimized for reversed-phase HPLC separations of peptides and polypeptides, using typical acidic mobile phases favored for proteomic applications.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 3/Pk.
pH 2 to 9
Particle Size 2.7 μm
Length (Metric) 0.5 cm
Pore Size 160 Å pore size
Surface Area 90 m2/g
Diameter (Metric) Inner 4.6 mm
For Use With (Equipment) HPLC: Suitable, L11 Per USP
Stationary Phase Phenyl-Hexyl (dimethylphenyl-hexylsilane) bonding
Endcapped Endcapped
Matrix Fused-Core™ particle platform
Show More Show Less

BIOshell is a trademark of Sigma-Aldrich Co. LLC, Fused-Core is a registered trademark of Advanced Materials Technology, Inc.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.