Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Analytics™ Biosystems Calibration Gas

Catalog No. 19806867
Encompass
Encompass_Preferred
Change view
Click to view available options
Concentration:
20.9%
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
19-806-867 20.9%
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-806-867 Supplier Honeywell Analytics™ Supplier No. 549039

Please call customer service for current pricing. 1-800-766-7000

missing translation for 'validMainframeMsg'

For ToxiPro and PHD6

  • Single component gas-O2 (zero air)(20.9%)
  • Balance N2
  • Five year shelf-life
  • 103L (27.21gallons)

TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 27.2 gal.
Capacity (Metric) 103 L
Concentration 20.9%
Components Measured Oxygen
Type Calibration Gas
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.