Learn More
Honeywell Analytics™ Biosystems MultiPro Sensors
Catalog No. 19175353
Encompass_Preferred
Catalog No. 19-175-353 Supplier Honeywell Analytics™ Supplier No. 544219
Requires
Biosystems MultiPro Multi-Gas Detector for LEL And Oxygen (Cat. No. 5448300A)
  • Field replaceable
  • Sensor span reserve indicators for predictive maintenance
TRUSTED_SUSTAINABILITY
Specifications
Type Gas Sensor
Sensor Types CO-H sensor