Description
- Streamlined Discovery: A single, comprehensive platform that covers a diverse set of biochemical and cell-based assays.
- Richer Cellular Insights: Capture more data per well with an expanded image cube capacity and enhanced optics.
- Optimized Workflow Speed: Experience faster imaging throughput to move from raw samples to analyzed results more efficiently.
- Superior Kinetic Data: Software-controlled gas modules create a stable, monitored environment for live cell studies.
- Ready for Any Assay: The modular design grows with your lab, allowing you to add imaging modes as needs evolve.
- This comprehensive system allows researchers to cover a diverse set of applications, from routine biochemical assays to complex cell-based studies.
Specifications
Specifications
| Type | Cell Imaging Multi-Mode Microplate Reader |
| Camera | Sony CMOS, 16-bit Gray Scale, Standard or WFOV |
| Includes | Automated Widefield Microscopy in Fluorescence, Phase Contrast, Brightfield, Color Brightfield, and High Contrast Brightfield. Variable Bandwidth Monochromator Detection of Fluorescence Intensity, UV-Vis Absorbance, and Luminescence. Incubation to 65°C and Shaking. |
| Model | MPW-SN |
| Magnification | 1.25x to 60x |