Learn More
Agilent BioTek Cytation 9 Cell Imaging Multimode Reader PROMO
GREENER_CHOICE
Catalog No. CYT9WSNP Shop All Agilent Technologies Products
Change view
Click to view available options
Model:
M-SN
MF-SN
MFA-SN
MFAW-SN
MFW-SN
MPW-SN
MW-SN
PW-SN
U-SN
UMW-SN
UW-SN
W-SN
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
CYT9WSNP W-SN
CYT9MFASNP MFA-SN
CYT9MFSNP MF-SN
CYT9MFAWSNP MFAW-SN
CYT9MFWSNP MFW-SN
CYT9MPWSNP MPW-SN
CYT9MSNP M-SN
CYT9USNP U-SN
CYT9UMWSNP UMW-SN
CYT9UWSNP UW-SN
CYT9MWSNP MW-SN
CYT9PWSNP PW-SN
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. CYT9WSNP Supplier Agilent Supplier No. CYT9WSN3416
Add to Cart
Add to Cart
It merges multimode microplate reading with widefield inverted and upright microscopy in a single platform.
  • Streamlined Discovery: A single, comprehensive platform that covers a diverse set of biochemical and cell-based assays.
  • Richer Cellular Insights: Capture more data per well with an expanded image cube capacity and enhanced optics.
  • Optimized Workflow Speed: Experience faster imaging throughput to move from raw samples to analyzed results more efficiently.
  • Superior Kinetic Data: Software-controlled gas modules create a stable, monitored environment for live cell studies.
  • Ready for Any Assay: The modular design grows with your lab, allowing you to add imaging modes as needs evolve.
  • This comprehensive system allows researchers to cover a diverse set of applications, from routine biochemical assays to complex cell-based studies.
TRUSTED_SUSTAINABILITY
Specifications
Type Cell Imaging Multi-Mode Microplate Reader
Camera Sony CMOS, 16-bit Gray Scale, Standard or WFOV
Includes Automated Widefield Microscopy in Fluorescence, Brightfield, Color Brightfield, and High Contrast Brightfield. Incubation to 65°C and Shaking.
Model W-SN
Magnification 1.25X to 60X