Learn More
Agilent BioTek Gas Controllers: Calibrated CO2 Gas Tank for CO2 Controller
Catalog No. BT01503 Shop All Agilent Technologies Products
Change view
Click to view available options
For Use With (Equipment):
CO2 Gas Controller
Catalog No. For Use With (Equipment)
BT01503 CO2 Gas Controller
1 options
This item is not returnable. View return policy
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. BT01503 Supplier Agilent BioTek Supplier No. 01503
Add to Cart
Add to Cart
This item is not returnable. View return policy
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Replacement tank for Accuracy Verification Kit
Calibrated CO2 gas tank, replacement for kit p/n 1210011 (does not include regulator)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Calibrated Carbon Dioxide Gas Tank
For Use With (Equipment) CO2 Gas Controller