Learn More
Agilent BioTek Gas Controllers: Calibration Kit for CO2 Controller
Shop All Agilent Technologies Products
Description
- Accuracy Verification Kit for both Gas Controllers (CO2 and O2 or CO2 only)
- Includes calibrated 5% CO2 tank, regulator and tubing
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Product Type | Calibration Kit |
| For Use With (Equipment) | CO2 Gas Controller |