BioTek™ MicroFlo™ Select Dispenser
Choose dynamic dispense volume, microplate types, tube sizes and dispense height
Manufacturer: BioTek™ 7170015
 View more versions of this product
Catalog No. 11-120-623
  Check Availability
Add to Cart
    Specifications
    Product Type Low Volume Cassette
    For Use With (Equipment) For el406, multiflo™, multiflo™ fx
    Volume (Metric) Range 1 to 50μL
    Diameter (English) 0.0075 in.
    Diameter (Metric) 0.019cm
    For Use With (Application) For liquid handling
    Tip Material Stainless Steel
    Dispensing Increments 1μL
  • Description & Specifications
Related Products
Product Content Feedback