Learn More
Apexbio Technology LLC Biotin-11-dCTP(Synonyms: Biotin-11-2'-deoxycytidine-5'-triphosphate, Biotin-11-dCTP trisodium salt), 1ml (1mM), CAS: 136632-30-9.
SDP
Supplier:  Apexbio Technology LLC B83461
Encompass_Preferred
Biotin-11-dCTP (CAS 136632-30-9) is a biotinylated nucleotide analog designed for incorporation into DNA or cDNA in enzymatic reactions such as PCR and nick translation It substitutes natural dCTP through polymerase-mediated incorporation The biotin moiety linked via an 11-atom spacer at cytidine s C5 position facilitates subsequent detection utilizing streptavidin-conjugated molecules (e g horseradish peroxidase alkaline phosphatase fluorescent dyes or affinity beads) Biotin-11-dCTP serves as an essential reagent to produce detectable nucleic acid probes commonly applied in molecular hybridization assays and genomic studies
Catalog No. 50-000-60497
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.