Learn More
Biolegend Biotin (azide-free) anti-β-Amyloid, 1-16
Supplier:  Biolegend 803009
Encompass_Preferred
Biotin (azide-free) anti-β-Amyloid, 1-16 [6E10]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: IHC-P; Size: 1 mL
Catalog No. 50-208-9879