Learn More
Tonbo Bioscience Antibodies Biotin Anti-Mouse CD19 (1D3) ..... Cat. No. 30-0193-U025 ..... Size 25 µg
Supplier:  Tonbo Bioscience 300193U025
Encompass_Preferred
Antibodies Biotin Anti-Mouse CD19 (1D3) ..... Cat. No. 30-0193-U025 ..... Size 25 µgThe 1D3 antibody reacts with mouse CD19, a 95 kDa glycoprotein which acts as a co-receptor, along with CD21 and CD81, in support of the functional B cell receptor (BCR). This complex provides antigen-specific recognition and subsequent activation of B cells to proliferate and differentiate into antibody-secreting cells (plasma cells) or memory B cells, which are crucial for secondary antigen encounter. CD19 is a lineage-differentiation marker, as its expression is detectable at the earliest B cell stages, through development, and is finally lost upon transition to mature plasma cells. The 1D3 antibody is widely used as a phenotypic marker for CD19 expression on B cells, as well as on dendritic cell subsets.
Catalog No. 50-210-3156
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.