Learn More
Cytek Biosciences Inc Antibodies Biotin Anti-Mouse CD252 (OX40 Ligand) (RM134L) ..... Cat. No. 30-5905-U025 ..... Size 25 µg
Supplier:  Cytek Biosciences Inc 305905U025
Encompass_Preferred
Antibodies Biotin Anti-Mouse CD252 (OX40 Ligand) (RM134L) ..... Cat. No. 30-5905-U025 ..... Size 25 µgThe RM134L antibody recognizes CD252, also known as OX-40 Ligand or CD134 Ligand, a member of the TNF-superfamily that is present on the surface of antigen presenting cells and activated B lymphocytes. The OX-40 Ligand interacts with OX-40 (CD134) which is expressed primarily on activated T cells. This costimulatory interaction leads to increased proliferation and IL-2 production responses of activated T cells, and at the same time enhances proliferation and immunoglobulin secretion by activated B cells.The RM134L antibody is useful for flow cytometric detection of CD252 on stimulated mouse splenocytes. It has also been reported to block the costimulatory activity of OX-40 Ligand. Please choose the appropriate format for each application.
Catalog No. 50-210-3180
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.