Learn More
CPC Scientific Biotin-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-OH (trifluoroacetate salt) 0.1MG
SDP
Supplier:  CPC Scientific AMYD010A
Encompass
beta-Amyloid (1-40) found to exhibit both neurotrophic and neurotoxic effects that depend on neuronal age and beta-protein concentration.ONE-LETTER SEQUENCE: Biotin-DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVMOLECULAR FORMULA: C204H309N55N60S2MOLECULAR WEIGHT:4556.2STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [183906-14-1], SYNONYMS: Biotinyl-Amyloid β-Protein (1-40)RESEARCH AREA: Alzheimer's DiseaseREFERENCES: B.A. Yankner et al., Science, 250, 279 (1990)
Catalog No. 50-210-8657
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.