Learn More
Description
Biotin Check&Go! is a simple immunochromatography test that requires only 10 minutes and small quantities of antibody conjugates.
The key components of the kit include a nitrocellulose membrane containing a Test line of immobilized Protein A and Protein G, both which have a high affinity for the Fc region of a variety of IgG molecules, and a 40 nm InnovaCoat GOLD Streptavidin conjugate for detection.
- Quickly check the successof your conjugation
- No costly equipment is required
- Compatible with antibody conjugates generated using our Lightning-Link™ Biotin antibody labeling kits and other biotin labeling technologies
Specifications
Specifications
| Target | Biotin Check&Go! |
| Content And Storage | Storage of components varies. See protocol for specific instructions. |
| Product Line | Novus |
| Quantity | 30 Tests |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
