Learn More
Bon Opus Biosciences Biotinylated Cynomolgus FGL1 (C-Avi-6His) -100ug
SDP
Supplier:  Bon Opus Biosciences CY20100UG
Encompass_Preferred
Recombinant Cynomolgus Fibrinogen-like Protein 1 is produced by our Mammalian expression system and the target gene encoding Leu23-Ile312 is expressed with a Avi, 6His tag at the C-terminus. Expression system: Human Cells. Accession# A0A2K5X990. Formulation: Supplied as a 0.2 μm filtered solution of 20mM Tris, 300mM Nacl, 10%Glycerol, 2mM EDTA, pH8.0. Bon Opus Cat# CY20
Catalog No. 50-208-0511
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.