Learn More
Bon Opus Biosciences Biotinylated Human CA125 (C-Fc-Avi) -20ug
SDP
Supplier:  Bon Opus Biosciences CY1020UG
Encompass_Preferred
Biotinylated Recombinant Human Mucin-16 is produced by our Mammalian expression system and thetarget gene encoding Gly12660-Met12923 is expressed with a Fc, Avi tag at the C-terminus. Expression system: Human Cells. Accession# Q8WXI7. Formulation: Lyophilized from a 0.2 μm filtered solution of 50 mM Tris, 100 mM Glycine, pH 7.5. Bon Opus Cat# CY10
Catalog No. 50-208-0494
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.