Learn More
Bon Opus Biosciences Biotinylated Recombinant Human EpCAM (C-6His-Avi) -20ug
SDP
Supplier:  Bon Opus Biosciences CY0320UG
Encompass_Preferred
Biotinylated Recombinant Human Epithelial Cell Adhesion Molecule is produced by our Mammalianexpression system and the target gene encoding Gln24-Lys265 is expressed with a 6His, Avitag at the C-terminus. Expression system: Human 293 Cells. Accession# AAH14785.1. Formulation: Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Bon Opus Cat# CY03
Catalog No. 50-208-0481
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.