Learn More
Bon Opus Biosciences Biotinylated Human IL-2RB (C-6His-Avi) -100ug
SDP
Supplier:  Bon Opus Biosciences CY31100UG
Encompass_Preferred
Biotinylated Recombinant Human Interleukin-2 Receptor subunit beta is produced by our Mammalian expression system and the target gene encoding Ala27-Asp239 is expressed with a 6His, Avi tag at the C-terminus. Expression system: Human Cells. Accession# P14784. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# CY31
Catalog No. 50-208-0527
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.