Learn More
Echelon Biosciences Research Labs Biotin Phosphatidylinositol 4-phosphate 50 ug
SDP
Supplier:  Echelon Biosciences Research Labs C04B650UG
Encompass
Biotin Phosphatidylinositol 4-phosphate is a water soluble analog of PI(4)P labeled with biotin at the sn-1 position.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 4-phosphate (PI(4)P) is the biosynthetic precursor to PI(4,5)P2 and has an important roles in regulating sphingomyelin and glycosphingolipid metabolism and membrane trafficking at the exit of the Golgi complex. This item is non returnable
Catalog No. 50-720-056
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.