Learn More
Description
PA1-30595 detects Biotin from goat samples.
Specifications
Specifications
| Antigen | Biotin |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | HRP |
| Formulation | 50mM HEPES with proprietary stabilizer and 0.002% thimerosal |
| Gene Alias | Vitamin B4 |
| Host Species | Goat |
| Immunogen | Goats were immunized with biotinylated protein carrier in Freunds adjuvant. |
| Purification Method | Antigen affinity chromatography |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
