Learn More
Genscript Corporation Biotin-Protein L
Supplier:  Genscript Corporation M00097
Encompass_Preferred
Protein L is a cell surface protein from Peptostreptoccocus magnus that binds to the variable light chains (kappa chain) of immunoglobulins without interfering with antigen binding. In contrast to IgG-binding proteins, such as protein A and protein G, protein L can be used for the detection and purification of mammalian kappa light chain antibodies of all classes. Biotin-Protein L, which is produced by covalently linking biotin to protein L, can be used in immunodetections, such as immunoblotting (western or dot blot), ELISA, and immunohistochemistry.
Catalog No. NC0988081
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.