Learn More
Apexbio Technology LLC Biotin-tyramide, 1g. Cas: 41994-02-9 MFCD: MFCD27953026. Reagent used for tyramide signal amplification (TSA)
SDP
Supplier:  Apexbio Technology LLC A80111000
Encompass_Preferred
Biotin-tyramide is used for tyramide signal amplification (TSA) which is a powerful, patented technology that significantly enhances both chromogenic and fluorescent signals. TSA is easily integrated into standard nonradioactive in situ hybridization (ISH) or IHC protocols Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4984
Due to product restrictions, please to purchase or view availability for this product.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.