Learn More
Enzo Life Sciences Ubiquitin, (biotinylated) (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLUW87050100
Encompass_Preferred
Biotinylated ubiquitin (prepared by the action of biotinamidocaproic acid N-hydroxysuccinimide ester on ubiquitin) is useful for studying ubiquitinylation in vitro, where it represents an alternative to radio-iodinated ubiquitin. This product contains a number of multiply biotinylated ubiquitin species, which has proven itself to be a reagent of considerable utility particularly for carrying out E1/E2-mediated thiolester assays. Formulation: Lyophilized from Acetonitrile:water (1:1). Appearance: White solid. Source: Produced in E. coli. Solubility: In order to ensure full solubility it is recommended that the material be dissolved in 5µL DMSO prior to dilution with the buffer of choice to the desired working. Concentration. UniProt: P0CG47 (UBB), P0CG48 (UBC), P62979 (RPS27A), P62987 (UBA52). Handling: Avoid freeze/thaw cycles. Long Term Storage: -20°C
Catalog No. NC9706714
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.