Learn More
Selleck Chemical LLC Biotin Vitamin B7-100mg
SDP
Supplier:  Selleck Chemical LLC S3130100MG
Encompass_Preferred
Biotin(Vitamin B7) is a water-soluble B-vitamin and acts as a necessary coenzyme for cell growth, the production of fatty acids, and the metabolism of fats and amino acids. Biotin is covalently attached to distinct lysine residues in histones, affecting chromatin structure and mediating gene regulation.
Catalog No. 50-238-9867
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.