Learn More
Bachem Biotinyl-Amyloid Beta Protein (1-42) Ammonium
SDP
Supplier:  Bachem 4038795.0500
Encompass_Preferred
Biotinyl-Amyloid Beta Protein (1-42) Ammonium, Biotinyl-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-Ile-Ala-OH, 1802086-20-9, C213H325N57O62S2, Mr 4740.40, 0.5mg. Biotinyl-b-Amyloid (1-42). N-terminally biotin-labeled Ab42.
Catalog No. 50-259-968