Learn More
Quidel Corporation monoclonal antibody to human C4d
SDP
Supplier:  Quidel Corporation A704
Encompass_Preferred
A biotin conjugated murine monoclonal antibody to human C4d containing fragments.
Catalog No. NC2000338
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.