Learn More
ACROBiosystems ACROBiosystems Biotinylated Anti-SARS-CoV-2 Spike RBD Antibody, Chimeric mAb, Human IgG1 (AM130)
Supplier:  ACROBiosystems S1NM13L3200UG
Encompass_Preferred
Antibody;200UG;Human IgG1/kappa;This product is a specific antibody against SARS-CoV-2 Spike RBD. No cross-reactivity is detected with Spike RBD of other coronaviruses, including SARS-CoV, MERS-CoV, HCoV-229E, HCoV-NL63, HCoV-OC43 and HCoV-HKU1.;Biotinylated Anti-SARS-CoV-2 Spike RBD Antibody, Chimeric mAb, Human IgG1 (AM130) is a chimeric monoclonal antibody recombinantly expressed from HEK293 cells, which combines the variable region of a mouse monoclonal antibody with human IgG1 constant domain. The mouse monoclonal antibody was obtained from a mouse immunized with recombinant SARS-CoV-2 S1 protein. This chimeric antibody is purified by Protein A affinity chromatography.; The protein has a calculated MW of 49.1 kDa | 23.5 kDa. The protein migrates as 27 kDa and 55 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Anti-SARS-CoV-2 Spike RBD Antibody, Chimeric mAb, Human IgG1 (AM122) (Cat. No.S1N-M12A1) at 2 μg/mL, add increasing concentrations of SARS-CoV-2 S protein RBD, His T
Catalog No. 50-210-7173
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.