Learn More
ACROBiosystems ACROBiosystems Biotinylated Cynomolgus LIV-1 / SLC39A6 Protein, His,Avitag™
Supplier:  ACROBiosystems LV1C82E525UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Cynomolgus LIV-1, His,Avitag (LV1-C82E5) is expressed from human 293 cells (HEK293). It contains AA Leu 21 - Ile 309 (Accession # XP_005586923.1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 36.2 kDa. The protein migrates as 50-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Cynomolgus LIV-1, His,Avitag (Cat. No. LV1-C82E5) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate can bind Anti-LIV-1 Antibody, Human IgG1 with a linear range of 0.1-4 ng/mL (QC tested).
Catalog No. 50-210-6350
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.