Learn More
ACROBiosystems ACROBiosystems Biotinylated Human ACE2 / ACEH Protein, His,Avitag™ (MALS verified)
Supplier:  ACROBiosystems AC2H82E7100MG
Encompass_Preferred
Recombinant Protein;100MG;Biotinylated Human ACE2, His,Avitag (AC2-H82E7) is expressed from human 293 cells (HEK293). It contains AA Gln 18 - Ser 740 (Accession # Q9BYF1-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 87.2 kDa. The protein migrates as 95-125 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized SARS-CoV-2 S1 protein, His Tag (Cat. No. S1N-C52H3) at 2 μg/mL (100 μL/well) can bind Biotinylated Human ACE2, His,Avitag (Cat. No. AC2-H82E7) with a linear range of 2-40 ng/mL (QC tested).
Catalog No. 50-210-7350
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.