Learn More
ACROBiosystems ACROBiosystems Biotinylated Human B7-H7 / HHLA2 Protein, Fc,Avitag™
Supplier:  ACROBiosystems B77H82F525UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human B7-H7, Fc,Avitag (B77-H82F5) is expressed from human 293 cells (HEK293). It contains AA Ile 23 - Asn 344 (Accession # Q9UM44-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 65.9 kDa. The protein migrates as 80-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD28H, Fc Tag (Cat. No. CDH-H5251) at 5 μg/mL (100 μL/well) can bind Biotinylated Human B7-H7, Fc,Avitag (Cat. No. B77-H82F5) with a linear range of 5-78 ng/mL (QC tested).
Catalog No. 50-210-6396
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.