Learn More
ACROBiosystems Biotinylated Human CA125 / MUC16 (13810-14451) Protein, Fc,Avitag™
Supplier: ACROBiosystems CA5H82F8200UG
Recombinant Protein;200UG;Biotinylated Human CA125 (13810-14451), Fc,Avitag (CA5-H82F8) is expressed from human 293 cells (HEK293). It contains AA Pro 13810 - Pro 14451 (Accession # Q8WXI7-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 100.0 kDa. The protein migrates as 120-160 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.