Learn More
ACROBiosystems ACROBiosystems Biotinylated Human Carbonic Anhydrase IX / CA9 (38-414) Protein, His,Avitag™ (MALS verified)
Supplier:  ACROBiosystems CA9H82E325UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human Carbonic Anhydrase IX (38-414), His,Avitag (CA9-H82E3) is expressed from human 293 cells (HEK293). It contains AA Gln 38 - Asp 414 (Accession # Q16790-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 44.5 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human Carbonic Anhydrase IX (38-414), His,Avitag (Cat. No. CA9-H82E3) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate can bind Anti-Carbonic Anhydrase IX Antibody, Human IgG1 with a linear range of 0.1-3 ng/mL (QC tested).
Catalog No. 50-210-6538
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.