Learn More
ACROBiosystems ACROBiosystems Biotinylated Human CD2 / SRBC Protein, His,Avitag™ (MALS verified)
Supplier:  ACROBiosystems CD2H82E6200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human CD2, His,Avitag (CD2-H82E6) is expressed from human 293 cells (HEK293). It contains AA Lys 25 - Asp 209 (Accession # AAH33583).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 24.8 kDa. The protein migrates as 35-43 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.;Immobilized Human CD58, Fc Tag (Cat. No. LF3-H5256) at 5 μg/mL (100 μL/well) can bind Biotinylated Human CD2, His,Avitag (Cat. No. CD2-H82E6) with a linear range of 0.2-1.25 μg/mL (QC tested).
Catalog No. 50-210-7129
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.