Learn More
ACROBiosystems ACROBiosystems Biotinylated Human CD5 Protein, Fc,Avitag™ (MALS verified)
Supplier:  ACROBiosystems CD5H82F325UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human CD5, Fc,Avitag (CD5-H82F3) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Pro 372 (Accession # P06127-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 66.8 kDa. The protein migrates as 80-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6211
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.