Learn More
ACROBiosystems ACROBiosystems Biotinylated Human CX3CL1 Protein, His Tag, ultra sensitivity (primary amine labeling)
Supplier:  ACROBiosystems CX1H8221200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human CX3CL1, His Tag, primary amine labeling (CX1-H8221) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - Gln 341 (Accession # AAH01163). It is the biotinylated form of Human CX3CL1, His Tag (Cat. No.CX1-H5221).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 35.5 kDa. The protein migrates as 50-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6777
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.