Learn More
ACROBiosystems ACROBiosystems Biotinylated Human FGF basic Protein, Avitag™,His Tag (MALS verified)
Supplier:  ACROBiosystems FGCH81E3200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human FGF basic, Avitag,His Tag (FGC-H81E3) is expressed from E.coli cells. It contains AA Pro 143 - Ser 288 (Accession # P09038-4).;This protein carries an Avi tag at the N-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 20.1 kDa. The protein migrates as 20-22 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Human Glypican 3, Fc Tag, low endotoxin (Cat. No. GP3-H5258) at 5 μg/mL (100 μL/well) can bind Biotinylated Human FGF basic, Avitag,His Tag (Cat. No. FGC-H81E3) with a linear range of 0.6-10 ng/mL (QC tested).;Immobilized Human FGFR2 (IIIb), His Tag (Cat. No. FGB-H5223) at 5 μg/mL (100 μL/well) can bind Biotinylated Human FGF basic, Avitag,His Tag (Cat. No. FGC-H81E3) with a linear range of 0.01-0.313 μg/mL (Routinely tested).
Catalog No. 50-210-6242
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.