Learn More
ACROBiosystems Biotinylated Human HLA-A*24:02&B2M&Survivin-2B (AYACNTSTL) Complex Protein (Monomer, MALS verified)
Supplier:  ACROBiosystems HLNH82E325UG
Encompass_Preferred
Survivin, also known as BIRC5 and EPR-1, is a member of the inhibitor of apoptosis (IAP) family, which encodes negative regulatory proteins that prevent apoptotic cell death. IAP family members usually contain multiple baculovirus IAP repeat (BIR) domains, but birc5 encodes proteins with only a single BIR domain. The Human HLA-A*2402 Survivin-2B (AYACNTSTL) complex protein is a complex of HLA-A*2402 of the MHC Class I, B2M and AYACNTSTL peptide of the Survivin-2B.
Catalog No. 50-271-9414
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.