Learn More
ACROBiosystems Biotinylated Human IL-3 R alpha / CD123 Protein, His,Avitag™ (MALS verified)
Supplier: ACROBiosystems ILAH82H525UG
Recombinant Protein;25UG;Biotinylated Human IL-3 R alpha, His,Avitag (ILA-H82H5) is expressed from human 293 cells (HEK293). It contains AA Thr 19 - Arg 305 (Accession # P26951-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 36.7 kDa. The protein migrates as 50-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human IL-3 R alpha, His,Avitag (Cat. No. ILA-H82H5) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate can bind Anti-IL-3 R alpha Antibody, Human IgG1 with a linear range of 0.4-2 ng/mL (QC tested).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.