Learn More
ACROBiosystems ACROBiosystems Biotinylated Human IL-3 R alpha / CD123 Protein, Fc,Avitag™
Supplier: ACROBiosystems ILAH82F325UG
Recombinant Protein;25UG;Biotinylated Human IL-3 R alpha, Fc,Avitag™ (ILA-H82F3) is expressed from human 293 cells (HEK293). It contains AA Thr 19 - Arg 305 (Accession # P26951-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 61.2 kDa.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.