Learn More
ACROBiosystems ACROBiosystems Biotinylated Human IL-8 protein, His,Avitag™
Supplier:  ACROBiosystems IL8H82H325UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human IL-8, His,Avitag (IL8-H82H3) is expressed from human 293 cells (HEK293). It contains AA Ser 28 - Ser 99 (Accession # P10145-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 12.0 kDa. The protein migrates as 14 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human IL-8, His,Avitag (Cat. No. IL8-H82H3) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate can bind Monoclonal Anti-Human IL-8 Antibody, Human IgG1 with a linear range of 0.06-2 ng/mL (QC tested).;Immobilized Monoclonal Anti-Human IL-8 Antibody, Human IgG1 at 1 μg/mL (100 μL/well) can bind Biotinylated Human IL-8, His,Avitag (Cat. No. IL8-H82H3) with a linear range of 0.06-0.2 ng/mL (Routinely tested).
Catalog No. 50-210-7395
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.