Learn More
ACROBiosystems ACROBiosystems Biotinylated Human Integrin isoform alpha-7X2B beta 1 (ITGA7X2B&ITGB1) Heterodimer Protein, Tag Free&His,Avitag™
Supplier:  ACROBiosystems IT1H82W525UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human ITGA7X2B&ITGB1 Heterodimer Protein, Tag Free&His,Avitag (IT1-H82W5) is expressed from human 293 cells (HEK293). It contains AA Phe 34 - Trp 1038 (ITGA7X2B) & Gln 21 - Asp 728 (ITGB1) (Accession # Q13683-7 (ITGA7X2B) & P05556-1 (ITGB1)).;Biotinylated Human ITGA7X2B & ITGB1 Heterodimer Protein, produced by co-expression of ITGA7X2B and ITGB1, has a calculated MW of 115.2 kDa (ITGA7X2B) and 86.7 kDa (ITGB1). Subunit ITGA7X2B is fused with an acidic tail but no tag at the C-terminus and subunit ITGB1 contains a basic tail at the C-terminus and followed by a polyhistidine tag and an Avi tag (Avitag™).
Catalog No. 50-210-6300
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.