Learn More
ACROBiosystems ACROBiosystems Biotinylated Human LILRB1 / CD85j / ILT2 Protein, Fc,Avitag™
Supplier:  ACROBiosystems CDJH82F7200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human LILRB1, Fc,Avitag (CDJ-H82F7) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - His 458 (Accession # D9IDM8-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 75.6 kDa. As a result of glycosylation, the protein migrates as 100-116 kDa under reducing (R) condition, and 150 kDa under non-reducing (NR) condition (SDS-PAGE).
Catalog No. 50-210-6585
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.