Learn More
ACROBiosystems ACROBiosystems Biotinylated Human LILRB4 / CD85k / ILT3 Protein, His,Avitag™ (MALS verified)
Supplier:  ACROBiosystems LI4H82E425UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human LILRB4, His,Avitag (LI4-H82E4) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Glu 259 (Accession # AAH26309.1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 29.8 kDa. The protein migrates as 32-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6288
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.