Learn More
ACROBiosystems Biotinylated Human MSP1D1 Protein, His,Avitag™ (Nanodisc)
Supplier:  ACROBiosystems APOH81Q5150UG
Encompass_Preferred
Recombinant Protein;150UG;Biotinylated Human MSP1D1, His,Avitag™ (Nanodisc) is expressed from E.coli cells. It contains AA Ser 1- Gln 211.; The protein has a calculated MW of 29.4 kDa. The protein migrates as 28 kDa under reducing (R) condition (SDS-PAGE).;2e5 of CD20-CAR-293 cells transfected with anti-CD20-scFv were stained with 100 μL of 10 μg/mL of Biotinylated Human MSP1D1 Protein, His,Avitag™ (Nanodisc) (Cat. No. APO-H81Q5), washed and then followed by PE-SA and analyzed with FACS. PE-SA was used as negative control (QC tested).
Catalog No. 50-205-5207
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.