Learn More
ACROBiosystems ACROBiosystems Biotinylated Human NCAM-1 / CD56 Protein, His,Avitag™ (MALS verified)
Supplier:  ACROBiosystems NC1H82E3200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human NCAM-1, His,Avitag (NC1-H82E3) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Gly 718 (Accession # P13591-2).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 81.0 kDa. The protein migrates as 95-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human NCAM-1, His,Avitag (Cat. No. NC1-H82E3) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate can bind Anti-NCAM-1 Antibody, Human IgG1 with a linear range of 1-5 ng/mL (QC tested).
Catalog No. 50-210-6610
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.