Learn More
ACROBiosystems Biotinylated Human PTK7 / CCK4 protein, His,Avitag™
Supplier:  ACROBiosystems PT7H82E525UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human PTK7, His,Avitag (PT7-H82E5) is expressed from human 293 cells (HEK293). It contains AA Ala 31 - Thr 704 (Accession # Q13308-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 78.2 kDa.
Catalog No. 50-205-4800
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.