Learn More
ACROBiosystems ACROBiosystems Biotinylated Human TIGIT Protein, Mouse IgG2a Fc,Avitag™ (MALS verified)
Supplier:  ACROBiosystems TITH82F325UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human TIGIT, Mouse IgG2a Fc,Avitag (TIT-H82F3) is expressed from human 293 cells (HEK293). It contains AA Met 22 - Pro 141 (Accession # Q495A1-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 41.6 kDa. The protein migrates as 45-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD155, Fc Tag (Cat. No. CD5-H5251) at 5 μg/mL (100 μL/well) can bind Biotinylated Human TIGIT, Mouse IgG2a Fc,Avitag (Cat. No. TIT-H82F3) with a linear range of 1-10 ng/mL (QC tested).;Immobilized Biotinylated Human TIGIT, Mouse IgG2a Fc,Avitag (Cat. No. TIT-H82F3) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate can bind Human TIGIT Antibody with a linear range of 2-16 ng/mL (Routinely tested).
Catalog No. 50-210-7222
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.