Learn More
ACROBiosystems ACROBiosystems Biotinylated Mouse Mesothelin / MSLN Protein, His,Avitag™
Supplier:  ACROBiosystems MSNM82E7200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Mouse Mesothelin, His,Avitag (MSN-M82E7) is expressed from human 293 cells (HEK293). It contains AA Asp 298 - Ser 600 (Accession # Q61468-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 37.7 kDa. The protein migrates as 45-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Mouse Mesothelin, His,Avitag (Cat. No. MSN-M82E7) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate can bind Monoclonal Anti-Human MSLN Antibody, Human IgG1 with a linear range of 10-78 ng/mL (QC tested).
Catalog No. 50-210-6262
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.