Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Biotinylated Recombinant Human VEGF 165 Protein
SDP

Catalog No. BT10543025 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BT10543025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BT10543025 Supplier R&D Systems™ Supplier No. BT10543025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Biotinylated via amines

When Recombinant Human VEGFR1/Flt-1 Fc Chimera Recombinant Human VEGF R1/Flt-1 Fc Chimera (Catalog # 321-FL ) is immobilized at 0.2 μg/mL (100 μL/well), Biotinylated Recombinant Human VEGF 165 (Catalog # BT10543) binds with an ED50 of 0.8-6.4 ng/mL.

Specifications

Accession Number NP_001165097.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 7422
Molecular Weight (g/mol) M.W.-observed: 22-27 kDa, under reducing conditions, M.W.-theoretical: 19 kDa
Quantity 25 μg
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived human VEGF protein Ala27-Arg191
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor
Conjugate Biotin
Reconstitution Reconstitute at 250 μg/mL in 4 mM HCl.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein VEGF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.