Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Biotinylated RNA Positive Control Oligonucleotides (Poly-A)

Catalog No. Q152P0100
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q152P0100 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. Q152P0100 Supplier Thermo Scientific™ Supplier No. Q152P.0100
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotinylated RNA Positive Control Oligonucleotides (Poly-A)

Biotinylated RNA Positive Control Oligonucleotides (Poly-A)

TRUSTED_SUSTAINABILITY

Specifications

Product Type Oligonucleotides
Content And Storage Store in refrigerator (2°C to 8°C).
Shipping Condition Wet Ice
Label or Dye Biotin
Quantity 1 mL
Product Line REMBRANDT

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.